Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Lysozyme Ab ELISA Kit

indirect solid phase ELISA for the quantitative measurement of IgG class autoantibodies against Lysozyme in Human Serum or Plasma

Manufacturer:  Abnova™ KA1090

Catalog No. 89-027-228


Add to cart

Specifications

Specifications

6.3 to 100U/mL
Lysozyme Ab ELISA Kit is an indirect solid phase ELISA for the quantitative measurement of IgG class autoantibodies against Lysozyme in human serum or plasma.
Colorimetric
6 Vials, 1.5mL combined calibrator with IgG class Anti-lysozyme, 2 vials x 1.5mL Anti-lysozyme controls, 1 vial x 20mL Sample Buffer, 1 vial x 15mL Enzyme Conjugate solution, 1 vial x 15mL TMB Substrate solution, 1 vial x 15mL stop solution, 1 96-well plate, 1 vial x 20mL wash solution
96 Assays
Store the kit at 4°C.
10μL
HRP
0.5U/mL
Quantification
Lysozyme Ab ELISA Kit
Plasma, Serum
96 Assays
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit