Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Magnesium Assay Kit
To quantify magnesium in a variety of biological samples, functional study
Supplier: Abnova™ KA0813
Includes:
- Magnesium assay buffer, 25mL
- Magnesium developer (lyophilized), 1 vial
- Magnesium enzyme mix (lyophilized), 1 vial
- Magnesium standard (150nmol/μL), 0.1mL
Specifications
2 to 15nm | |
Magnesium Assay Kit is used for quantitating magnesium in a variety of biological samples. | |
Functional Study | |
1 Kit | |
Detection Assay Kit |
Store the kit at -20°C. | |
Colorimetric | |
25mL Magnesium assay buffer, 1 vial Magnesium developer (lyophilized), 1 vial Magnesium enzyme mix (lyophilized), 0.1mL Magnesium standard (150nM/μL) | |
Biological Fluid, Cell Lysate, Culture Medium, Serum, Tissue Lysate, Urine |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction