Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Maltose Assay Kit

Used for quantifying maltose and functional study

Manufacturer:  Abnova™ KA0851

Catalog No. 89-024-406


Due to product restrictions, please to purchase or view availability for this product.


Includes:
  • Maltose assay buffer, 25mL
  • Maltose probe (in DMSO), 200µL
  • α-D-Glucosidase (lyophilized), 1 vial
  • Enzyme mix (lyophilized), 1 vial
  • Maltose standard (100nmol/µL), 100µL

Specifications

Specifications

Maltose Assay Kit is used for quantifying maltose.
25mL Maltose assay buffer, 200μL Maltose assay buffer, 1 vial a-D-Glucosidase (lyophilized), 1 vial enzyme mix (lyophilized), 100μL Maltose standard (100nM/μL)
Biological fluid, cell culture supernatant, plasma, serum, tissue culture supernatant, urine
Colorimetric, Fluorometric
Functional Study
1 Kit
Store the kit at -20°C.
Maltose Assay Kit
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit