Learn More
Abnova™ Maltose and Glucose Assay Kit
To measure glucose or maltose levels, functional study
Manufacturer:  Abnova™ KA0843
Catalog No. 89-028-061
Add to cart
Specifications
Specifications
10pM to 10nM
Functional Study
1 Kit
Store the kit at -20°C.
Maltose and Glucose Assay Kit
Maltose and Glucose Assay Kit is used for measuring glucose or maltose levels.
25mL Glucose assay buffer, 1 vial Glucose probe, 0.4mL DMSO, 1 vial alpha-D-Glucosidase, 1 vial Glucose enzyme mix, 100μL Maltose standard
Biological fluid, cell culture supernatant, plasma, serum, tissue culture supernatant, urine
Fluorometric