Learn More
Abnova™ MBD2 Binding Activity/Inhibition Assay Kit
Measuring the binding activity of MBD2 to Methylated DNA
Manufacturer:  Abnova™ KA0675
Catalog No. 89-027-034
Add to cart
Specifications
Specifications
HRP
Colorimetric
25mL MB1 (Wash Buffer 10X), 3mL MB2 (Assay Buffer), 100μL MB3 (Methylated DNA 25μg/mL), 50μL MB4 (affinity Antibody 100μg/mL), 20μL MB5 (detection Antibody 200
MBD2 binding activity/inhibition assay kit
Store MB3, MB4, MB5, MB6, and 8 well strips at 4°C away from light. Store other components at room temperature.
MBD2 Binding Activity/Inhibition Assay Kit is use for measuring the binding activity of MBD2 to methylated DNA.
Functional Study
MBD2 Binding Activity/Inhibition Assay Kit
1 Kit
96 Assays