Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Methamphetamine ELISA Kit

Specific and sensitive in-vitro test to detect the presence of d-methamphetamine in samples such as whole blood, oral fluids, serum, plasma and urine

Manufacturer:  Abnova™ KA0934

Catalog No. 89-024-475


Due to product restrictions, please to purchase or view availability for this product.


Description

Description

  • Sample Type: Oral Fluid, Plasma, Serum, Urine, Whole Blood
  • Sample Volume: 10µL

Specifications

Specifications

10 to 50ng/mL
Methamphetamine ELISA Kit is a specific and sensitive in-vitro test to detect the presence of d-methamphetamine in samples such as whole blood, oral fluids, serum, plasma and urine.
Qualitative
Methamphetamine ELISA Kit
Oral Fluid, Plasma, Serum, Urine, Whole Blood
10μL
HRP
Colorimetric
Microwell strip 12 x 8 x 1, 12mL Enzyme Conjugate, 2mL positive reference standard, 1mL negative control, 12mL TMB chromogenic Substrate, 11mL stop solution
96 Assays
Store the kit at 4°C.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit