Learn More
Abnova™ Mitochondria/Cytosol Fractionation Kit
Mitochondria/Cytosol Fractionation Kit is used for isolation of a highly enriched mitochondrial fraction from cytosolic fraction of mammalian cells including both apoptotic and non-apoptotic cells.
Supplier:  Abnova™ KA0777
Catalog No. 89-028-003
Add to Cart
Description
Description
  • Provides unique formulations of reagents for effective isolation of a highly enriched mitochondrial fraction from cytosolic fraction of mammalian cells including both apoptotic and nonapoptotic cells
  • The enriched mitochondrial and cytosolic fractions can be used for studying apoptotic and signal transduction pathways to detect translocation of factors interested between the two fractions by Western blotting, ELISA, or other assays
  • Procedures are simple and easy to perform, no ultracentrifugations and toxic chemicals are involved
  • After opening the kit, store buffers at 4°C; store Protease Inhibitor Cocktail and DTT at -20°C
  • This kit has been configured for research use only and is not for diagnostic and clinical use
Specifications
Specifications
Mitochondria/Cytosol Fractionation Kit is used for isolation of a highly enriched mitochondrial fraction from cytosolic fraction of mammalian cells including both apoptotic and non-apoptotic cells.
1 Year
Functional Study
1 Kit
Store the kit at -20°C
Liquid (Triton X-100), Powder (PMSF), Liquid (Glycerol), Powder (DTT)
Product Suggestions
Product Suggestions