Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Mitochondrial DNA Isolation Kit
Mitochondrial DNA Isolation Kit is used for isolating mtDNA from a variety of cells and tissues.
Supplier: Abnova™ KA0895
Description
- The Mitochondrial DNA Isolation Kit provides convenient tools for isolating mtDNA from a variety of cells and tissues in high yield and purity, without contaminations from genomic DNA
- The purified mtDNA can be used for a variety of studies such as enzyme manipulations, Southern blotting, cloning, PCR analysis, and amplifications
- After opening the kit, store Enzyme B Mix at -70°C; store all other Buffers at 4°C
- Protect from light and store Incubation Buffer at 4°C after opening
- Stable for up to 3 months when properly stored
- This kit has been configured for research use only and is not for diagnostic and clinical use
Specifications
Mitochondrial DNA Isolation Kit is used for isolating mtDNA from a variety of cells and tissues. | |
DNA Isolation Kit | |
Functional Study |
20mL (5X) Cyto sol extraction Buffer, 1.8mL Mito chondrial Lysis Buffer, Enzyme B mix (lyophilized) 1Vial, 1.5mL TE Buffer | |
Store at -20°C | |
Each |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction