Learn More
Abnova™ Morphine (Canine) ELISA Kit
Specific and sensitive in-vitro test to detect the presence of Morphine in cCanine samples such as whole Blood, Serum, Plasma and Urine
Manufacturer:  Abnova™ KA0936
Catalog No. 89-024-477
Due to product restrictions, please to purchase or view availability for this product.
Specifications
Specifications
5 to 25ng/mL
Dog
1ng/mL
Qualitative
Morphine (Canine) ELISA Kit
Plasma, Serum, Urine, Whole Blood
20μL
HRP
Morphine (Canine) Specific ELISA Kit is a specific and sensitive in-vitro test to detect the presence of morphine in canine samples such as whole blood, serum, plasma and urine.
Colorimetric
Microwell strip 12 x 8 x 1, 12.5mL Enzyme Conjugate,1mL positive reference standard, 1mL negative control,14mL TMB chromogenic Substrate, 12.5mL stop solution
96 Tests
Store the kit at 4°C.