Abnova™ NAD+/NADH Assay Kit

Manufacturer: Abnova Corporation KA3798

 View more versions of this product

Catalog No. 89-101-286

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    For Use With (Application) Sensitive measurement of NAD, NADH and thrie ratio in cell or tissue extracts
    Quantity 1 Kit
    Sample Type Cell or Tissue Extract
    Sample Volume 25μL
    Storage Requirements Store the kit at -80°C.
    Detection Method Colorimetric

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit