Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Nickel Assay Kit
To quantify nickel, functional study
Supplier: Abnova™ KA0822
Specifications
2 to 50nmol | |
Nickel Assay Kit is used for quantifying Ni+2. | |
Functional Study | |
1 Kit | |
Detection Assay Kit |
Store the kit at room temperature. | |
Colorimetric | |
20mL Nickel buffer, 1mL Nickel reagent, 1 vial Nickel standard | |
Biological Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Culture Supernatant, Urine |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction