Learn More
Abnova™ Nitrate/Nitrite Colorimetric Assay Kit (LDH method)
A measurement for detecting low levels of nitrite. The kit is particularly well suited to measurements of NOS activity in vitro.
Manufacturer:  Abnova™ KA1343
Catalog No. 89-024-625
Add to cart
Description
Description
  • High throughput screening applications using recombinant NOS preparations can be quickly and accurately assayed with the LDH method kit
  • This kit can also be used for the assay of nitrate and nitrite in urine, plasma, serum, and tissue culture medium
Specifications
Specifications
Nitrate/Nitrite Colorimetric Assay Kit (LDH method) is a measurement for detecting low levels of nitrite.
1 Vial Nitrate/Nitrite Assay Buffer, 1 vial Nitrate Reductase (LDH method), 1 vial Lactate Dehydrogenase Co-factor preparation, 1 vial Nitrate standard, 1 vial Lactate Dehydrogenase, 1 vial Griess reagent R1, 1 vial Griess reagent R2, 1 plate 96-well plate (colorimetric assay), 1 cover 96-well cover sheets
Store the kit at -20°C temporarily. For best results, store the components as described in the protocol.
Nitrate/Nitrite Colorimetric Assay Kit (LDH method)
Functional Study
1 Kit
Colorimetric