Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Nitric Oxide Assay Kit (Fluorometric)
To measure total nitrate/nitrite concentration, functional study
Supplier: Abnova™ KA0793
Specifications
Store the kit at -20°C. | |
Fluorometric | |
30mL assay buffer, enzyme Co-factor (lyophilized), enhancer (lyophilized), nitrate Reductase (lyophilized), nitrate standard (lyophilized), nitrite standard (lyophilized), 1mL DAN probe, 10mL Sodium Hydroxide, 2 each plate cover, 2 microtiter plate | |
Biological Fluid, Cell Lysate, Culture Medium, Serum, Tissue Lysate, Urine |
Nitric Oxide Assay Kit (Fluorometric) is used to measuring total nitrate/nitrite concentration. | |
Functional Study | |
1 Kit | |
Detection Assay Kit |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction