Learn More
Abnova™ Opiates ELISA Kit
Specific and sensitive in-vitro test to detect the presence of Opiates in samples such as whole Blood, oral fluids, Serum, Plasma and Urine
Manufacturer:  Abnova™ KA0937
Catalog No. 89-024-478
Due to product restrictions, please to purchase or view availability for this product.
Specifications
Specifications
5 to 25ng/mL
Opiates ELISA Kit is a specific and sensitive in-vitro test to detect the presence of opiates in samples such as whole blood, oral fluids, serum, plasma and urine.
Colorimetric
Microwell strip 12 x 8 x 1, 12.5mL Enzyme Conjugate, 1mL positive reference standard, 1mL negative control, 14mL TMB chromogenic Substrate, 12.5mL stop solution
96 Assays
Store the kit at 4°C.
HRP
0.25ng/mL
Qualitative
Opiates ELISA Kit
Oral Fluid, Plasma, Serum, Urine, Whole Blood
10μL