Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Ovarian Ab ELISA Kit

Sandwich Enzyme Immunoassay for the quantitative determination of Anti-ovary antibodies in Human Serum

Manufacturer:  Abnova™ KA1260

Catalog No. 89-025-099


Add to cart

Specifications

Specifications

156 to 10000pg/mL
Human
78pg/mL
Quantification
Ovarian Ab ELISA Kit
1 Kit
Store the kit at 4°C.
HRP
Anti-Ovarian Ab ELISA Kit is a solid-phase sandwich enzyme immunoassay for the quantitative determination of anti-ovary antibodies in human serum.
Colorimetric
Ovarian Antibody ELISA standard set 4 vials, 0.5mL Control 1 vial, 0.5mL Dilution Buffer 1 vial, 50mL washing solution 1 vial, 50mL Enzyme Conjugate 1 vial, 5mL Substrate solution 1 vial, 13mL stop solution 1 vial, 13mL 96-well plate 1, Holder for single
Ovarian Ab ELISA kit
Serum
50μL
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit