Learn More
Abnova™ Oxaloacetate Assay Kit
Used for quantifying OAA and functional study
Manufacturer:  Abnova™ KA0867
 View more versions of this product
Catalog No. 89-024-422
Due to product restrictions, please to purchase or view availability for this product.
Specifications
Specifications
0.1 to 10nm
Functional Study
1 Kit
Store the kit at -20°C.
Oxaloacetate Assay Kit
Oxaloacetate Assay Kit is used for quantifying OAA.
25mL OAA assay buffer, 0.2mL OAA probe (in DMSO), OAA enzyme mix (lyophilized), Developer (lyophilized), OAA standard (10μM lyophilized)
Biological fluid, cell culture supernatant, plasma, serum, tissue culture supernatant, urine
Colorimetric, Fluorometric