Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Phosphate Assay Kit (Colorimetric)

To assess phosphate over a wide range of concentrations, functional study

Manufacturer:  Abnova™ KA0815

Catalog No. 89-028-084


Add to cart

Includes:
  • Phosphate reagent, 15mL
  • Phosphate standard (10mM), 0.5mL

Specifications

Specifications

1 to 1000μM
Functional Study
1 Kit
Store the kit at room temperature.
Phosphate Assay Kit
Phosphate Assay Kit (Colorimetric) is used for assessing phosphate over a wide range of concentrations.
15mL Phosphate reagent, 0.5mL Phosphate standard (10mM)
Biological fluid, cell lysate, culture medium, plasma, serum, tissue lysate, urine
Colorimetric
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit