Learn More
Abnova™ Phosphatidic Acid IgG/IgM ELISA Kit
Indirect Solid Phase ELISA for the quantitative measurement of IgG and IgM class autoantibodies against Phosphatidic Acid in Human Serum or Plasma
Manufacturer:  Abnova™ KA1269
Catalog No. 89-024-561
Add to cart
Specifications
Specifications
HRP
Anti-Phosphatidic Acid Ab IgG/IgM is an indirect solid phase enzyme immunoassay for the quantitative measurement of IgG and IgM class autoantibodies against phosphatidic acid in human serum or plasma.
Quantification
Phosphatidic Acid IgG/IgM ELISA Kit
1 Kit
Store the kit at 4°C.
Human
Colorimetric
1.5mL Combined calibrator with IgG and Ig class Anti-Phospholipid Antibodies 6 vials, 1.5mL Anti-Phospholipid Controls 2 vials, 20mL Sample Buffer 1 vial, 15mL Enzyme Conjugate solution 1 vial, 15mL Enzyme Conjugate solution 1 vial, 15mL TMB Substrate solution 1 vial, 96-well plate 1, 15mL stop solution 1 vial, 20mL Wash solution 1 vial
Phosphatidic Acid IgG/IgM ELISA kit
Plasma, Serum
100μL