Learn More
Abnova™ Phosphatidyl Serine Ab ELISA Kit
Indirect Solid Phase Enzyme Immunoassay for quantitative measurement of IgG and IgM class autoantibodies against Phosphatidyl Serine in Human Serum, Plasma
Manufacturer:  Abnova™ KA1277
Catalog No. 89-024-569
Due to product restrictions, please to purchase or view availability for this product.
Specifications
Specifications
HRP
Phosphatidyl Serine Ab ELISA Kit is an indirect solid phase enzyme immunoassay for the quantitative measurement of IgG and IgM class autoantibodies against phosphatidyl serine in human serum or plasma.
Quantification
Phosphatidyl Serine Ab ELISA Kit
1 Kit
Store the kit at 4°C.
Human
Colorimetric
1.5mL Combined calibrator with IgG and IgM class Anti-phospholipid Antibodies 6 vials, 1,5mL Anti-C1q controls in a Serum/Buffer Matrix 2 vials, 1,5mL Anti-Phospholipid Controls in a Serum/Buffer Matrix 2 vials, 20mL Sample Buffer 1 vial, 15mL Enzyme Conjugate solution 1 vial, 15mL TMB Substrate solution 1 vial, 15mL stop solution 1 vial, 96-well plate 1, 20mL Wash solution 1 vial
Phosphatidyl Serine Ab ELISA kit
Plasma, Serum
100μL