Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Phosphatidylcholine Assay Kit

Used for measuring phosphatidylcholine

Manufacturer:  Abnova™ KA0827

Catalog No. 89-028-073


Add to cart

Description

Description

  • Sample Type: Biological Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Culture Supernatant, Urine

Functional Study

Specifications

Specifications

0.1 to 10nm
Functional Study
1 Kit
Store the kit at -20°C.
Phosphatidylcholine Assay Kit
Phosphatidylcholine Assay Kit is used for measuring phosphatidylcholine.
25mL PC assay buffer, 0.2mL OxiRed™ probe, PC Hydrolysis enzyme (lyophilized), PC development mix (lyophilized), PC standard (10μM) (lyophilized)
Biological fluid, cell culture supernatant, plasma, serum, tissue culture supernatant, urine
Colorimetric, Fluorometric
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit