Learn More
Abnova™ PLTP Inhibitor Drug Screening Kit
To screen PLTP inhibitors, functional study
Manufacturer:  Abnova™ KA0830
Catalog No. 89-028-070
Add to cart
Specifications
Specifications
PLTP Activity Assay Kit can be used for screening PLTP inhibitors.
1mL PLTP donor molecule, 1mL PLTP acceptor molecule, 5mL PLTP assay buffer 10X, 0.3mL positive control (Rabbit Serum)
Biological fluid, cell culture supernatant, plasma, serum, tissue culture supernatant, urine
Fluorometric
Functional Study
1 Kit
Store the kit at 4°C.
PLTP Activity Assay Kit