Learn More
Abnova™ Proteasome Activity Assay Kit
Used for assaying the activity of proteasome and functional study
Manufacturer:  Abnova™ KA1431
Catalog No. 89-024-551
Due to product restrictions, please to purchase or view availability for this product.
Specifications
Specifications
Proteasome Activity Assay Kit is a fluorescence-based method for assaying the activity of proteasome.
25mL proteasome assay buffer, 100μL proteasome substrate (Succ-LLVY-AMC in DMSO), 100μL proteasome inhibitor (MG-132 in DMSO), 100μL AMC standard (1mM in DMSO), 1 vial positive control (lyophilized)
Cell lysate, cytoplasmic lysate, nuclear lysate
Store the kit at -20°C away from light.
Proteasome Activity Assay Kit
Functional Study
1 Kit
50μL
Fluorometric