Learn More
Abnova™ Protein Expression Kit I (GST-Fusion Protein)
To produce GST-fusion proteins at high purities
Manufacturer:  Abnova™ KA1435
Catalog No. 89-028-188
Add to cart
Specifications
Specifications
This kit produces GST-fusion proteins at high purities. The wheat germ included in this kit is a specialized wheat germ extract prepared by removing endogenous proteins as nonspecifically adsorb to GST-tag affinity purification resin.
2mL WEPRO™ 1240/WEPRO™ 1240H/WEPRO™ 1240G, 1.1mL SUB-AMIX™ S-1, 1.1mL SUB-AMIX™ S-2, 1.1mL SUB-AMIX™ S-3, 1.1mL SUB-AMIX™ S-4, 0.
Store the component at -20°C or -80°C as shown on the protocol.
To produce GST-fusion proteins at high purities
1 Kit
Protein Expression Kit I (GST-fusion Protein)