Learn More
Abnova™ Protein Expression Kit I
Used for general purpose, automatic or manual, protein synthesis using in-vitro wheat germ system
Manufacturer:  Abnova™ KA1434
Catalog No. 89-028-187
Add to cart
Specifications
Specifications
The protein expression kit I is used for general purpose, automatic or manual, protein synthesis using in vitro wheat germ system.
1mL WEPRO™ 1240/WEPRO™ 1240H/WEPRO™ 1240G, 1.1mL SUB-AMIX™ S-1, 1.1mL SUB-AMIX™ S-2, 1.1mL SUB-AMIX™ S-3, 1.1mL SUB-AMIX™ S-4, 0.
Store the component at -20°C or -80°C as shown on the protocol.
Used for general purpose, automatic or manual, protein synthesis using in-vitro wheat germ system
1 Kit
Protein Expression Kit I