Learn More
Abnova™ Protein Expression Kit II
The protein expression kit II is used for general purpose, automatic or manual, protein synthesis using
Manufacturer:  Abnova™ KA1437
Catalog No. 89-028-190
Add to cart
Description
Description
  • WEPRO7240 Expression Kit contains WEPRO™ 7240; suitable for protein expression where affinity purification to high purities is neither necessary nor desirable
  • WEPRO7240H Expression Kit is suitable for His-tagged protein expression and purification
  • WEPRO7240G Expression Kit is suitable for GST-tagged protein expression and purification
  • For research use only
Specifications
Specifications
The protein expression kit II is used for general purpose, automatic or manual, protein synthesis using in vitro wheat germ system.
Store the component at -20°C or -80°C as shown on the protocol.
1 Kit
For general purpose, auto matic or manual, Protein synthesis using in vitro wheat germ system
Protein Expression Kit II