Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ PTX3 (Human) ELISA Kit

Sandwich Enzyme Immunoassay for the quantitative measurement of Human PTX3

Manufacturer:  Abnova™ KA1007

 View more versions of this product

Catalog No. 89-028-131


Add to cart

Specifications

Specifications

0.5 to 20ng/mL
Human
0.1ng/mL
Quantification
PTX3 (Human) ELISA Kit
1 Kit
Cell Culture Supernatant, Plasma
96 Assays
HRP
PTX3 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of PTX3.
Colorimetric
96-well Antibody coated microwell, 12mL HRP Conjugated Anti-PTX3 Antibody reagent, 0.1mL x 7 vials Human PTX3 standard (0, 0.5, 1.0, 2.5, 5.0, 10, 20ng/mL), 100mL Wash Buffer 10X concentrate, 15mL Dilution Buffer, 12mL TMB solution, 12mL stop solution
PTX3 (Human) ELISA kit
RUO
Store the kit at 4°C.
10μL
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit