Learn More
Abnova™ Pyruvate Assay Kit
To measure pyruvate concentration, functional study
Manufacturer:  Abnova™ KA0836
Catalog No. 89-028-065
Add to cart
Specifications
Specifications
1μM to 10mM
Functional Study
1 Kit
Store the kit at -20°C.
Pyruvate Assay Kit
Pyruvate Assay Kit is used for measuring pyruvate concentration.
25mL Pyruvate assay buffer, 1 vial OxiRed™ probe, 400μL DMSO (anhydrous), 1 vial Pyruvate enzyme mix, 100μL Pyruvate standard
Biological fluid, cell culture supernatant, plasma, serum, tissue culture supernatant, urine
Colorimetric, Fluorometric