Learn More
Abnova™ RELA ELISA Kit
Sandwich Enzyme Immunoassay for the quantitative measurement of free RELA in the nucleus of cells or tissues
Manufacturer:  Abnova™ KA1394
Catalog No. 89-024-514
Due to product restrictions, please to purchase or view availability for this product.
Specifications
Specifications
Alkaline Phosphatase
RELA ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of free RELA in the nucleus of cells or tissues.
Quantification
RELA ELISA Kit
1 Kit
Store the standard and pNPP at -20°C. Store all other components at 4°C.
Human,Mouse,Rat
Colorimetric
200μL Capture Antibody, 200μL detecting Antibody, 2 x 10mL Coating Buffer, 2 x 0.5g BSA, 50mL Wash Buffer 20X, 10μL AKP Conjugated Secondary Ab, 2 x 10mL pNPP Substrate Buffer, 2 vials, lyophilized (0.
RELA ELISA kit
Cell Lysate, Tissue Lysate
100μL