Learn More
Abnova™ ReliableDAB Peroxidase Substrate Kit
Overcomes this disadvantage and can remain ready-to-use for 2 weeks after preparation
Manufacturer:  Abnova™ KA0562
Catalog No. 89-027-126
Add to cart
Description
Description
Immunohistochemistry
Specifications
Specifications
ReliableDAB Peroxidase Substrate Kits ReliableDAB Peroxidase Substrate kit overcomes this disadvantage and can remain ready-to-use for 2 weeks after preparation.
2mL Stable DAB solution, 100mL Buffer solution
ReliableDAB Peroxidase Substrate kit
Store the kit at 4°C.
Immunohistochemistry
ReliableDAB Peroxidase Substrate Kit
500 Slides