Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Rib-P Ab ELISA Kit

Indirect Solid Phase ELISA for the quantitative measurement of IgG class autoantibodies against ribosomal P Proteins in Human Serum or Plasma

Manufacturer:  Abnova™ KA1110

Catalog No. 89-027-248


Add to cart

Specifications

Specifications

12.5 to 200U/mL
Rib-P is an indirect solid phase ELISA for the quantitative measurement of IgG class autoantibodies against ribosomal P proteins in human serum or plasma.
Colorimetric
6 Vials, 1.5mL each Anti-Rib-P calibrator, 2 vials, 1.5mL each Anti-Rib-P Controls, 1 vial, 20mL Sample Buffer, 1 vial, 15mL Enzyme Conjugate solution, 1 vial, 15mL TMB Substrate solution, 1 plate 96-well plate, 1 vial, 15mL stop solution, 1 vial, 20mL wash solution
1 Kit
Store the kit at 4°C.
HRP
1U/mL
Quantification
Rib-P Ab ELISA Kit
Plasma, Serum
10μL
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit