Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Rubella virus IgG ELISA Kit

solid phase Enzyme Immunoassay for the quantitative measurement of IgG class autoantibodies against Rubella virus.

Manufacturer:  Abnova™ KA1458

 View more versions of this product

Catalog No. 89-028-298

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.


Specifications

Specifications

HRP
Colorimetric
96-well ELISA break away strip coated with specific Antigen, 1.2mL negative Control, 1.2mL/ each vial calibrator (10, 20, 100 and 200 IU), 12mL Antigen/Px Conjugate, 80mL Wash Buffer concentrate, 100mL Dilution Buffer, 13mL Chromogenic Substrate, 15mL stop solution
Rubella Virus IgG ELISA kit
RUO
Store at 4°C on dry atmosphere away from light.
100μL
Rubella virus IgG ELISA Kit is a solid phase enzyme immunoassay for the quantitative measurement of IgG class autoantibodies against Rubella virus.
Quantification
Rubella virus IgG ELISA Kit
1 Kit
Serum
96 Assays
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit