Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Rubella virus IgM ELISA Kit
solid phase Enzyme Immunoassay for the qualitative determination of IgM autoantibodies against Rubella virus.
Supplier: Abnova™ KA1459
Specifications
HRP | |
Colorimetric | |
1 Kit | |
Serum | |
Qualitative | |
96 |
Rubella virus IgM ELISA Kit is a solid phase enzyme immunoassay for the determination of IgM autoantibodies against Rubella virus. | |
96-well ELISA break away strip coated with specific Antigen, 1.2mL negative Control, 1.2mL positive Control, 1.2mL Cut-off Control, 12mL Antigen/Px Conjugate, 80mL Wash Buffer concentrate, 100mL Dilution Buffer, 13mL Chromogenic Substrate, 15mL stop solution | |
For research use only (RUO) | |
Store at 4°C on dry atmosphere away from light. | |
Rubella Virus IgM ELISA kit |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction