Learn More
Abnova™ Sarcosine Assay Kit
Used for measuring sarcosine and functional study
Manufacturer:  Abnova™ KA0857
Catalog No. 89-024-412
Due to product restrictions, please to purchase or view availability for this product.
Specifications
Specifications
1 to 10000μM
Functional Study
1 Kit
Store the kit at -20°C.
Sarcosine Assay Kit
Sarcosine Assay Kit is used for measuring sarcosine.
25mL Sarcosine assay buffer, 1vial Sarcosine probe, 0.4mL DMSO, 1 vial Sarcosine enzyme mix, 1 vial Sarcosine standard
Biological fluid, cell culture supernatant, plasma, serum, tissue culture supernatant
Colorimetric, Fluorometric