Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Serum Antibody Purification Kit

For measuring Trypsin activity using fluorometric methods

Manufacturer:  Abnova™ KA1596

Catalog No. 89-028-279


Add to cart

Specifications

Specifications

The Serum Antibody Purification Kit is used to purify IgG fractions from both serum and ascites fluid of antibodies.
Store at 4°C.
1 Kit
Purification
Serum Antibody Purification Kit
1 Vial Serum Protein A Resin, 1 vial 10X Binding Buffer, 1 vial Wash Buffer, 1 vial Elution Buffer, 1 vial neutraliser, 1 purification column
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit