Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Spermatozoa Ab ELISA Kit

Sandwich Enzyme Immunoassay for the quantitative determination of Anti-spermatozoa antibodies in Human seminal Plasma

Manufacturer:  Abnova™ KA1257

Catalog No. 89-025-096


Add to cart

Specifications

Specifications

0 to 60U/mL
Human
Colorimetric
4 Vial, 0.5mL Sperm Antibody ELISA standard set, 1 vial, 0.5mL Control, 1 vials, 50mL Dilution Buffer, 1 vials, 50mL Washing solution, 1 vial, 8mL Enzyme Conjugate, 1 vial, 13mL Substrate solution, 1 vial, 13mL stop solution, 1 plate 96-well plate, 1 x holder for single strip
Spermatozoa Ab ELISA kit
Seminal Plasma
50μL
HRP
Anti-Spermatozoa Ab ELISA Kit is a solid-phase sandwich enzyme-immunoassay for the quantitative determination of anti-spermatozoa antibodies in human seminal plasma.
Quantification
Spermatozoa Ab ELISA Kit
1 Kit
Store the kit at 4°C.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit