Learn More
Abnova™ Starch Assay Kit
Provides an easy, convenient method to measure starch levels in a variety of samples.
Manufacturer:  Abnova™ KA0862
Catalog No. 89-024-417
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
In the assay, starch is hydrolyzed to glucose which is oxidized to generate color and fluorescence. The assay can detect starch at 0.0004 to 2mg/mL
Specifications
Specifications
0.0004 to 2mg/mL
Functional Study
1 Kit
Store the kit at -20°C.
Starch Assay Kit
Starch Assay Kit is used for measuring starch levels.
25mL Hydrolysis buffer, 25mL Hydrolysis buffer, 1 vial OxiRed™ probe, 0.4mL DMSO (anhydrous), 1 vial Hydrolysis enzyme mix, 1 vial Development enzyme mix, 0.1mL Starch standard
Biological fluid, cell culture supernatant, plasma, serum, tissue culture supernatant, urine
Colorimetric, Fluorometric