Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Superoxide Dismutase Activity Assay Kit Available on GSA/VA Contract for Federal Government customers only.

Used for detecting SOD activity.

Manufacturer:  Abnova™ KA0783

Catalog No. 89-028-110


Add to cart

Includes:
  • WST Solution, 1mL
  • SOD Enzyme solution, 20µL
  • SOD Assay buffer, 20mL
  • SOD Dilution buffer, 10mL

Description

Description

  • The sensitive SOD assay kit utilizes WST-1, which produces a water-soluble formazan dye upon reduction with superoxide anion
  • The rate of the reduction with a superoxide anion is linearly related to the xanthine oxidase (XO) activity, and is inhibited by SOD. Therefore, the inhibition activity of SOD can be determined by a colorimetric method

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Superoxide Dismutase Activity Assay Kit is used for detecting SOD activity (% inhibition rate).
1mL WST solution, 20μL SOD enzyme solution, 20mL SOD assay buffer, 10mL SOD Dilution buffer
Biological fluid, cell lysate, culture medium, plasma, serum, tissue lysate, urine
Colorimetric
Functional Study
100 Assays
Store the kit at 4°C.
Superoxide Dismutase Activity Assay Kit
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit