Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ TCS Antibody Purification Kit

For measuring Trypsin activity using fluorometric methods

Manufacturer:  Abnova™ KA1597

Catalog No. 89-028-280


Add to cart

Specifications

Specifications

The TCS Antibody Purification Kit is used to purify IgG fractions from hybridoma supernatants of antibodies.
Store at 4°C.
1 Kit
Purification
TCS Antibody Purification Kit
1 Vial TCS Protein A Resin, 1 vial 10X Binding Buffer, 1 vial Wash Buffer, 1 vial Elution Buffer, 1 vial neutraliser
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit