Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Thioredoxin Reductase Assay Kit

Thioredoxin Reductase Assay Kit is used for detecting TrxR activity.

Manufacturer:  Abnova™ KA0883

Catalog No. 89-024-438


Due to product restrictions, please to purchase or view availability for this product.


Includes:
  • TrxR assay buffer, 25mL
  • TNB standard (lyophilized), 1 vial
  • DTNB (lyophilized), 1 vial
  • NADPH (lyophilized), 1 vial
  • TrxR positive control, 1 vial
  • TrxR inhibitor (lyophilized), 1 vial

Description

Description

  • Kit provides a convenient colorimetric assay for detecting TrxR activity in various samples
  • Since in crude biological samples other enzymes, such as glutathione reductase and glutathione peroxidase, can also reduce DTNB, therefore, TrxR specific inhibitor is utilized to determine TrxR specific activity
  • Two assays are performed: the first measurement is of the total DTNB reduction by the sample, and the second one is the DTNB reduction by the sample in the presence of the TrxR specific inhibitor
  • The difference between the two results is the DTNB reduction by TrxR
  • Store the kit at -20°C, protect from light
  • For research use only

Specifications

Specifications

Thioredoxin Reductase Assay Kit is used for detecting TrxR activity.
DTNB (lyophilized) 1 vial
Store the kit at -20°C
100 Assays
Functional Study
Thioredoxin Reductase Assay Kit
100 Assays
Biological Fluids, Cell Culture Supernatant, Plasma, Serum, Tissue Culture Supernatant, Urine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit