Learn More
Abnova™ Tissue Methyl-CpG Binding domain Protein 2 ChIP Kit
Tissue Methyl-CpG Binding domain Protein 2 ChIP Kit is for investigating the interactions of the in vivo binding of MBD2 to methylated DNA in a variety of mammalian tissues.
Manufacturer:  Abnova™ KA0695
Catalog No. 89-027-052
Add to cart
Description
Description
  • The fastest procedure available, which can be finished within 5 hours
  • Strip microplate format makes the assay flexible: manual or high throughput
  • Columns for DNA purification are included: save time and reduce labor
  • Compatible with all DNA amplification-based approaches
  • Simple, reliable, and consistent assay conditions
  • Upon receipt store the following components at 4°C: Protease inhibitor cocktail, normal mouse IgG, Anti-MBD2, proteinase K, control primers, 8-well assay strips; store all other components at room temperature
  • The kit is stable for up to 6 months from the shipment date, when stored properly
  • This kit has been configured for research use only and is not for diagnostic and clinical use
Specifications
Specifications
Tissue Methyl-CpG Binding domain Protein 2 ChIP Kit is for investigating the interactions of the in vivo binding of MBD2 to methylated DNA in a variety of mammalian tissues.
Tissue Methyl-CpG Binding domain Protein 2 ChIP Kit
Store the following components at 4°C: Protease inhibitor cocktail, normal mouse IgG, Anti-MBD2, proteinase K, 8 well strips. Store other components at room temperature.
Chromatin Immunoprecipitation
1 Kit