Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Total Antioxidant Capacity Assay Kit

To measure the combination of both small molecule antioxidants and proteins and small molecules alone in the presence of our proprietary protein mask, functional study

Manufacturer:  Abnova™ KA0802

Catalog No. 89-028-094


Add to cart

Includes:
  • Cu2+ Reagent, 0.2mL
  • Assay diluent, 10mL
  • Protein mask, 10mL
  • Trolox standard (lyophilized), 1µmol

Specifications

Specifications

Total Antioxidant Capacity Assay Kit is used for measuring the combination of both small molecule antioxidants and proteins and small molecules alone in the presence of our proprietary Protein Mask.
0.2mL Cu2+ reagent, 10mL assay diluent, 10mL protein mask, 1 vial trolox standard (1μM)
Biological fluid, cell lysate, culture medium, plasma, serum, tissue lysate, urine
Colorimetric
Functional Study
1 Kit
Store the kit at 4°C.
Total Antioxidant Capacity Assay Kit
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit