Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Total Histone H3 Acetylation Detection Fast Kit (Colorimetric)

Total Histone H3 Acetylation Detection Fast Kit (Colorimetric) is used for measuring total acetylation of histone H3.

Manufacturer:  Abnova™ KA1538

Catalog No. 89-028-232


Add to cart

Description

Description

  • Quick and efficient procedure, which can be finished within 2.5 hours
  • Innovative colorimetric assay without the need for radioactivity, electrophoresis, and chromatography
  • Captures histone H3 acetylated at any lysine site with the detection limit as low 2 ng/well and detection range from 5 ng-2 µg/well of histone extracts
  • The control is conveniently included for the quantification of the amount of acetylated histone H3
  • Strip microplate format makes the assay flexible: manual or high throughput
  • Simple, reliable, and consistent assay conditions
  • Upon receipt store C3 and the Standard Control at -20°C; store all other components at 4°C away from light
  • The kit is stable for up to 6 months from the shipment date, when stored properly
  • For research use only

Specifications

Specifications

Total Histone H3 Acetylation Detection Fast Kit (Colorimetric) is used for measuring total acetylation of histone H3.
Total Histone H3 Acetylation Detection Fast Kit (Colorimetric)
Histone extracts from mammalian cells/ tissues
1 Kit
Functional Study
50 μL
Store the detection antibody and the standard at -20°C. Store all other components at 4°C away from light.
Histone Extract
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit