Learn More
Abnova™ Toxoplasma gondii IgA ELISA Kit
Indirect Solid Phase ELISA for the determination of IgA autoantibodies against Toxoplasma gondii
Manufacturer:  Abnova™ KA1462
Catalog No. 89-028-302
Add to cart
Specifications
Specifications
HRP
Colorimetric
96-well ELISA break away strip coated with specific Antigen 96-well, 1.2mL negative control, 1.2mL positive control, 1.2mL Cut off control, 12mL Antigen/Px Conjugate, 80mL Wash Buffer concentrate, 100mL Dilution Buffer, 13mL Chromogenic Substrate, 15mL stop solution
Toxoplasma gondii IgA ELISA kit
Serum
100μL
Toxoplasma gondii IgA ELISA Kit is a solid phase enzyme immunoassay for the determination of IgA autoantibodies against Toxoplasma gondii.
Qualitative
Toxoplasma gondii IgA ELISA Kit
1 Kit
Store at 4°C on dry atmosphere away from light.