Learn More
Abnova™ Treponema pallidum IgM ELISA Kit
For the detection of IgM antibody to Treponema pallidum in Human Serum or Plasma
Manufacturer:  Abnova™ KA0969
 View more versions of this product
Catalog No. 89-028-050
Add to cart
Specifications
Specifications
HRP
Colorimetric
Microwell 12 x 8 x 1, 22mL sample diluent, 1.5mL calibrator, 1.5mL positive control, 1.5mL negative control, 12mL Enzyme Conjugate, 12mL TMB chromogenic Substrate, 12mL stop solution, 25mL Washing concentrate (20X)
96 Assays
Plasma, Serum
96 Assays
Treponema pallidum IgM ELISA Kit is used for the detection of IgM antibody to Treponema pallidum in human serum or plasma.
Qualitative
Treponema pallidum IgM ELISA Kit
RUO
Store the kit at 4°C.
10μL