Learn More
Abnova™ TRF (Rabbit) ELISA Kit
highly sensitive two-site enzyme-linked immunoassay for the quantitative determination of TRF in Rabbit Serum or Plasma
Manufacturer:  Abnova™ KA1977
Catalog No. 89-028-692
Add to cart
Specifications
Specifications
3.12 to 100ng/mL
Rabbit
Colorimetric
50mL Diluent concentrate, 50mL wash solution concentrate, 150μL Enzyme Antibody Conjugate concentrate, 12mL TMB Substrate solution, 12mL stop solution, 96-well microtiter plate, calibrator (lyophilized) 1 vial
TRF (Rabbit) ELISA kit
Plasma, Serum
100μL
HRP
TRF (Rabbit) ELISA Kit is a highly sensitive two-site enzyme-linked immunoassay for the quantitative determination of TRF in rabbit serum or plasma.
Quantification
TRF (Rabbit) ELISA Kit
1 Kit
Store the kit at 4°C.