Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Triglyceride Quantification Kit
Click to view available options
Quantity:
1 Kit
Description
- Sample Type: Biological Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Culture Supernatant, Urine
Functional Study
Specifications
Specifications
Calibration Range | 2 to 10000μM |
Content And Storage | Store the kit at -20°C. |
Description | Triglyceride Quantification Kit is used for measuring TG concentration. |
Detection Method | Colorimetric, Fluorometric |
For Use With (Application) | Functional Study |
Includes | 25mL Triglyceride assay buffer, 200μL Triglyceride probe (in DMSO, anhydrous), 1 vial Lipase, 1 vial Triglyceride enzyme mix (lyophilized), 0.3mL Triglyceride standard (1mM) |
Quantity | 1 Kit |
Sample Type | Biological Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Culture Supernatant, Urine |
Type | Quantification Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction