Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Trypsin Activity Assay Kit

For measuring Trypsin activity using fluorometric methods

Manufacturer:  Abnova™ KA0779

Catalog No. 89-028-001


Add to cart

Includes:
  • Trypsin assay buffer, 25mL
  • Trypsin Substrate (in DMSO), 200µL
  • Positive Control (~2U, Lyophilized), 1 vial.
  • p-NA Standard (2mM), 400µL
  • Trypsin Inhibitor (TLCK, 20mM), 100µL
  • Chymotrypsin Inhibitor (TPCK, 10mM), 100µL

Specifications

Specifications

Mammals
Colorimetric
25mL Trypsin Assay Buffer, 200μL Trypsin Substrate, positive control 1 vial, 400μL p-NA standard, 100μL Trypsin inhibitor, 100μL Chymotrypsin inhibitor
1 Kit
Store the kit at -20°C.
Trypsin Activity Assay Kit is use for measuring trypsin activity using fluorometric methods.
Functional Study
Trypsin activity assay kit
Biological Fluid, Cell Culture Supernatant, Plasma, Serum, Tissue Culture Supernatant, Urine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit