Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Xanthine Oxidase Assay Kit

Used for determining XO and functional study

Manufacturer:  Abnova™ KA0874

Catalog No. 89-024-429


Due to product restrictions, please to purchase or view availability for this product.


Includes:
  • XO Assay Buffer 25mL
  • OxiRed Probe (in DMSO) 200µL
  • XO Enzyme Mix lyophilized
  • XO Substrate Mix lyophilized
  • XO Positive Control µL
  • H2O2 Standard (0.88M) 0.1mL

Specifications

Specifications

1 to 100mU
Functional Study
1 Kit
Store the kit at -20°C.
Xanthine Oxidase Assay Kit
Xanthine Oxidase Assay Kit is used for determining XO.
25mL XO assay buffer, 200μL OxiRed™ probe (in DMSO), 1 vial XO enzyme mix (lyophilized), 1 vial XO substrate mix (lyophilized), 8μL XO positive control, 0.1mL H2O2 standard (0.88M)
Biological fluid, cell culture supernatant, plasma, serum, tissue culture supernatant, urine, food sample, growth medium
Colorimetric, Fluorometric
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit