Learn More
Abnova™ Zinc Assay Kit
Manufacturer:  Abnova Corporation KA3706
 View more versions of this product
Catalog No. 89-101-225
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Colorimetric
Quantitative colorimetric zinc determination at 425 nm
1 Kit
Store the kit at 4°C.
Zinc Assay Kit
50mL Reagent A, 1mL reagent B, 1mL reagent C,1mL (100mM) EDTA, 1mL (50μM) Zinc standard
Biological Fluid, Cell Lysate, Cerebrospinal Fluid, Culture Medium, Plasma, Serum, Tissue Lysate, Urine