Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ ABpure™ Mouse Antibody Purification Kit
SDP

Novus Biologicals™ ABpure Mouse Antibody Purification Kit is used to remove contaminants such as BSA and other proteins, glycine, tris, and azide that interfere in conjugation reactions.

$542.50 - $1026.00

Specifications

Content And Storage 4°C
Product Type Antibody Purification Kit
View More Specs

Alert:

Not suitable for use with antibodies from other species


Products 2
Catalog Number Mfr. No. Sufficient For Price Quantity & Availability  
Catalog Number Mfr. No. Sufficient For Price Quantity & Availability  
26-100-10
SDP
View Documents
Novus Biologicals™
2610010
Three purifications
Each for $1,026.00
Add to Cart
 
26-100-05
SDP
View Documents
Novus Biologicals™
2610005
One purification
Each for $542.50
Add to Cart
 
Description

Description

  • Used to purify antibodies from crude samples such as ascites fluid or immune serum
  • Antibody to be purified or cleaned up ideally is in volume of 0.1mL to 0.5mL
  • 20 to 150μg of antibody can be purified in each run
Specifications

Specifications

4°C
Antibody Purification Kit
SDS
Documents

Documents

Product Certifications

For research use only.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit